Stampa…
Anti-DAB1 Rabbit polyclonal antibody unconjugated
Fornitore: Abcam
ABCAAB111684-100EA 895 EUR
ABCAAB111684-100
Anti-DAB1 Rabbit polyclonal antibody unconjugated
Anticorpi
Type: Primary
Antigen: DAB1 (Dab, reelin signal transducer, homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse