Stampa…
Anti-FLNA Rabbit polyclonal antibody unconjugated
Fornitore: Abcam
ABCAAB111620-100EA 878 EUR
ABCAAB111620-100
Anti-FLNA Rabbit polyclonal antibody unconjugated
Anticorpi
Type: Primary
Antigen: FLNA (filamin A, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human