Anti-PLA2R Goat Polyclonal Antibody
ABCAAB99466-100
: Abcam
- Antibody type:Primary
- Antigen symbol:PLA2R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pack type:Vial
- Pk:100 µG