Stampa…
Anti-WIF1 Mouse Monoclonal Antibody [clone: MM0602-6D27]
Fornitore: Abcam
ABCAAB89935-50EA 882 EUR
ABCAAB89935-50
Anti-WIF1 Mouse Monoclonal Antibody [clone: MM0602-6D27]
Anticorpi
Type: Primary
Antigen: WIF1 (WNT inhibitory factor 1)
Clonality: monoclonal
Clone: MM0602-6D27
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human