Stampa…
Anti-VCAN Mouse monoclonal antibody unconjugated [clone: MM0600-7D41]
ABCAAB89934-50EA 666 EUR
ABCAAB89934-50
Anti-VCAN Mouse monoclonal antibody unconjugated [clone: MM0600-7D41]
Type: Primary
Antigen: VCAN (versican)
Clonality: monoclonal
Clone: MM0600-7D41
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity:
Visualizza i codici