Stampa…
Anti-MCSF Receptor Mouse Monoclonal Antibody [clone: MM0467-5V36]
Fornitore: Abcam
ABCAAB89907-50EA 916 EUR
ABCAAB89907-50
Anti-MCSF Receptor Mouse Monoclonal Antibody [clone: MM0467-5V36]
Anticorpi
Type: Primary
Antigen: MCSF Receptor
Clonality: monoclonal
Clone: MM0467-5V36
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human