Stampa…

Anti-LIFR Mouse Monoclonal Antibody [clone: MM0455-9B23]

Fornitore: Abcam
ABCAAB89792-50EA 876 EUR
ABCAAB89792-50
Anti-LIFR Mouse Monoclonal Antibody [clone: MM0455-9B23]
Anticorpi
Type: Primary
Antigen: LIFR (leukemia inhibitory factor receptor alpha)
Clonality: monoclonal
Clone: MM0455-9B23
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR