Stampa…
Anti-FTH1 Mouse Polyclonal Antibody
Fornitore: Abcam
ABCAAB89787-50EA 882 EUR
ABCAAB89787-50
Anti-FTH1 Mouse Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: FTH1 (ferritin, heavy polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human