Stampa…
Anti-PCCA Mouse Polyclonal Antibody
Fornitore: Abcam
ABCAAB89784-50EA 882 EUR
ABCAAB89784-50
Anti-PCCA Mouse Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: PCCA (propionyl CoA carboxylase, alpha polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human