Stampa…
Anti-SOST Mouse Monoclonal Antibody [clone: MM0556-9N13]
Fornitore: Abcam
ABCAAB89733-50EA 876 EUR
ABCAAB89733-50
Anti-SOST Mouse Monoclonal Antibody [clone: MM0556-9N13]
Anticorpi
Type: Primary
Antigen: SOST (sclerostin)
Clonality: monoclonal
Clone: MM0556-9N13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human