Stampa…
Anti-RNase H2 subunit C Rabbit Polyclonal Antibody
Fornitore: Abcam
ABCAAB89726-100EA 896 EUR
ABCAAB89726-100
Anti-RNase H2 subunit C Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: RNase H2 subunit C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human