Stampa…
Anti-Retinol dehydrogenase 16 Rabbit Polyclonal Antibody
Fornitore: Abcam
ABCAAB89653-200EA 657 EUR
ABCAAB89653-200
Anti-Retinol dehydrogenase 16 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Retinol dehydrogenase 16
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human