Stampa…
Anti-ACAT2 Rabbit Polyclonal Antibody
Fornitore: Abcam
ABCAAB74901-100EA 838 EUR
ABCAAB74901-100
Anti-ACAT2 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: ACAT2 (acetyl-CoA acetyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human