Stampa…

Anti-EPHA2 Rat Monoclonal Antibody [clone: RM-0051-8F21]

Fornitore: Abcam
ABCAAB73254-100EA 913 EUR
ABCAAB73254-100
Anti-EPHA2 Rat Monoclonal Antibody [clone: RM-0051-8F21]
Anticorpi
Type: Primary
Antigen: EPHA2 (EPH receptor A2)
Clonality: monoclonal
Clone: RM-0051-8F21
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR