Stampa…
Anti-PADI3 / PAD3 Rabbit Polyclonal Antibody
Fornitore: Abcam
ABCAAB50246-200EA 877 EUR
ABCAAB50246-200
Anti-PADI3 / PAD3 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: PADI3 / PAD3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat