Stampa…

Anti-HDAC7 Mouse monoclonal antibody unconjugated [clone: HDAC7-97]

ABCAAB50212-100EA 688 EUR
ABCAAB50212-100
Anti-HDAC7 Mouse monoclonal antibody unconjugated [clone: HDAC7-97]
Type: Primary
Antigen: HDAC7 (histone deacetylase 7)
Clonality: monoclonal
Clone: HDAC7-97
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR