Stampa…

Anti-GUSB Rabbit polyclonal antibody unconjugated

ABCAAB50148-100EA 666 EUR
ABCAAB50148-100
Anti-GUSB Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: GUSB (glucuronidase, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR