Anti-M13 phage coat protein g8p Mouse Monoclonal Antibody
BIRBORB98316-0.1
: Biorbyt
- Antibody type:Primary
- Antigen name:M13 phage coat Protein g8p
- Antigen symbol:M13
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Western blot:Yes
- Storage buffer:PBS containing 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:0,1 mg
About this item
Type: Primary
Antigen: M13 phage coat protein g8p
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: