Stampa…

Anti-CASP3 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB126549-100EA 0 EUR
BIRBORB126549-100 BIRBORB126549-200
Anti-CASP3 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: CASP3 (caspase 3, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR