Stampa…

Anti-Caspase 1 (p10) Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB126544-200EA 0 EUR
BIRBORB126544-200 BIRBORB126544-100
Anti-Caspase 1 (p10) Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Caspase 1 (p10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR