Stampa…

Anti-Cytochrome P450 11B1/2 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB126541-200EA 0 EUR
BIRBORB126541-200 BIRBORB126541-100
Anti-Cytochrome P450 11B1/2 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Cytochrome P450 11B1/2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR