Stampa…

Anti-RNF113B Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB126539-200EA 0 EUR
BIRBORB126539-200 BIRBORB126539-100
Anti-RNF113B Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: RNF113B (ring finger protein 113B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR