Stampa…

Anti-PKCB1 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB126504-100EA 0 EUR
BIRBORB126504-100 BIRBORB126504-200
Anti-PKCB1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: PKCB1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR