Stampa…

Anti-ZC3H11A Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB126497-100EA 0 EUR
BIRBORB126497-100 BIRBORB126497-200
Anti-ZC3H11A Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: ZC3H11A (zinc finger CCCH-type containing 11A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR