Stampa…
Anti-ZAR1 Rabbit Polyclonal Antibody
Fornitore: Biorbyt
BIRBORB126496-200EA 0 EUR
BIRBORB126496-200 BIRBORB126496-100
Anti-ZAR1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: ZAR1 (zygote arrest 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human