Stampa…

Anti-XRCC5 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB126492-200EA 0 EUR
BIRBORB126492-200 BIRBORB126492-100
Anti-XRCC5 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: XRCC5 (X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR