Stampa…

Anti-XRCC4 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB126491-100EA 0 EUR
BIRBORB126491-100 BIRBORB126491-200
Anti-XRCC4 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: XRCC4 (X-ray repair complementing defective repair in Chinese hamster cells 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR