Stampa…

Anti-SAA4 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB100586-200EA 0 EUR
BIRBORB100586-200 BIRBORB100586-100
Anti-SAA4 Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; IC 1:100-500; Immunofluorescence: 1:50-200

Type: Primary
Antigen: SAA4 (serum amyloid A4, constitutive)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR