Stampa…
Anti-PRDM2 Rabbit Polyclonal Antibody
Fornitore: Biorbyt
BIRBORB100514-100EA 0 EUR
BIRBORB100514-100 BIRBORB100514-200
Anti-PRDM2 Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: PRDM2 (PR domain containing 2, with ZNF domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat