Stampa…

Anti-CST6 Rabbit polyclonal antibody

Fornitore: Biorbyt
BIRBORB100512-200EA 0 EUR
BIRBORB100512-200 BIRBORB100512-100
Anti-CST6 Rabbit polyclonal antibody
Anticorpi
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: CST6 (cystatin E/M)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR