Stampa…

Anti-ARID4B Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB100507-100EA 0 EUR
BIRBORB100507-100 BIRBORB100507-200
Anti-ARID4B Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: ARID4B (AT rich interactive domain 4B (RBP1-like))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR