Stampa…

Anti-Ribonuclease 6 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB100504-100EA 0 EUR
BIRBORB100504-100 BIRBORB100504-200
Anti-Ribonuclease 6 Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; IC 1:100-500; Immunofluorescence: 1:50-200

Type: Primary
Antigen: Ribonuclease 6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR