Stampa…

Anti-BCL2L15 Rabbit polyclonal antibody

Fornitore: Biorbyt
BIRBORB100406-100EA 0 EUR
BIRBORB100406-100 BIRBORB100406-200
Anti-BCL2L15 Rabbit polyclonal antibody
Anticorpi
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: BCL2L15 (BCL2-like 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR