Stampa…

Anti-HPN Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB100345-100EA 0 EUR
BIRBORB100345-100 BIRBORB100345-200
Anti-HPN Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: HPN (hepsin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR