Stampa…

Anti-CD200 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB100118-100EA 0 EUR
BIRBORB100118-100 BIRBORB100118-200
Anti-CD200 Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: CD200 (CD200 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR