Stampa…

Anti-EZH2 Rabbit polyclonal antibody

Fornitore: Biorbyt
BIRBORB66896-100EA 0 EUR
BIRBORB66896-100
Anti-EZH2 Rabbit polyclonal antibody
Anticorpi
Type: Primary
Antigen: EZH2 (enhancer of zeste homolog 2 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR