Stampa…

Anti-IL5 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB66883-100EA 0 EUR
BIRBORB66883-100
Anti-IL5 Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:1000-1:10000

Type: Primary
Antigen: IL5 (interleukin 5 (colony-stimulating factor, eosinophil))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR