Stampa…

Anti-CDH2 Rabbit polyclonal antibody

Fornitore: Biorbyt
BIRBORB66835-100EA 0 EUR
BIRBORB66835-100
Anti-CDH2 Rabbit polyclonal antibody
Anticorpi
  • Western Blot: 1:1000-1:5000

Type: Primary
Antigen: CDH2 (cadherin 2, type 1, N-cadherin (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR