Stampa…

Anti-CCL3 Rabbit polyclonal antibody

Fornitore: Biorbyt
BIRBORB66825-100EA 0 EUR
BIRBORB66825-100
Anti-CCL3 Rabbit polyclonal antibody
Anticorpi
ELISA: 1:1000-1:10000

Type: Primary
Antigen: CCL3 (chemokine (C-C motif) ligand 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR