Stampa…

Anti-CSF2 Rabbit polyclonal antibody

Fornitore: Biorbyt
BIRBORB66817-100EA 0 EUR
BIRBORB66817-100
Anti-CSF2 Rabbit polyclonal antibody
Anticorpi
Western Blot: 1:1000-1:5000; ELISA: 1:1000-1:10000

Type: Primary
Antigen: CSF2 (colony stimulating factor 2 (granulocyte-macrophage))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR