Stampa…

Anti-IL33 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB66786-100EA 0 EUR
BIRBORB66786-100
Anti-IL33 Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:1000-1:5000; ELISA: 1:1000-1:10000

Type: Primary
Antigen: IL33 (interleukin 33)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR