Stampa…

Anti-RBBP5 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB66744-100EA 0 EUR
BIRBORB66744-100
Anti-RBBP5 Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: RBBP5 (retinoblastoma binding protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR