Stampa…

Anti-PIAS1 Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB66686-100EA 0 EUR
BIRBORB66686-100
Anti-PIAS1 Mouse Monoclonal Antibody
Anticorpi
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: PIAS1 (protein inhibitor of activated STAT, 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR