Stampa…

Anti-PECAM1 Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB66672-100EA 0 EUR
BIRBORB66672-100
Anti-PECAM1 Mouse Monoclonal Antibody
Anticorpi
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR