Stampa…

Anti-CBR1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornitore: Biorbyt
BIRBORB66478-100EA 0 EUR
BIRBORB66478-100
Anti-CBR1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Western Blot: 1-1000

Type: Primary
Antigen: CBR1 (carbonyl reductase 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR