Stampa…

Anti-ATXN3 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB49146-100EA 0 EUR
BIRBORB49146-100 BIRBORB49146-200
Anti-ATXN3 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: ATXN3 (ataxin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR