Stampa…

Anti-FEN1 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB49073-200EA 0 EUR
BIRBORB49073-200 BIRBORB49073-100
Anti-FEN1 Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1500: 12000; Immunochemistry: 150: 1200

Type: Primary
Antigen: FEN1 (flap structure-specific endonuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR