Stampa…

Anti-CDC25A Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB49071-100EA 0 EUR
BIRBORB49071-100 BIRBORB49071-200
Anti-CDC25A Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: CDC25A (cell division cycle 25A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR