Stampa…

Anti-FBL Rabbit polyclonal antibody

Fornitore: Biorbyt
BIRBORB49031-100EA 0 EUR
BIRBORB49031-100 BIRBORB49031-200
Anti-FBL Rabbit polyclonal antibody
Anticorpi
Type: Primary
Antigen: FBL (fibrillarin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR