Stampa…

Anti-AMACR Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB49023-100EA 0 EUR
BIRBORB49023-100 BIRBORB49023-200
Anti-AMACR Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1500: 12000; Immunochemistry: 150: 1200

Type: Primary
Antigen: AMACR (alpha-methylacyl-CoA racemase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR